RIE156322475 Rieke Metals, Inc.

3-Chloro-4-pyridyl 1-propylbutyl ketone

  • MDL number MFCD07699456
  • Cas Number : 898785-73-4
  • Molecular Weight : 239.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-20
Availability details for
Quantity
  • Availability
  • Add to Cart