RIE156322476 Rieke Metals, Inc.

2-chloro-5-(5-methoxycarbonyl-2-furoyl)pyridine

  • MDL number MFCD07699458
  • Cas Number : 898785-77-8
  • Molecular Weight : 265.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5575

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5575
Availability details for
Quantity
  • Availability
  • Add to Cart