RIE156322478 Rieke Metals, Inc.

Methyl 2-(2-methoxynicotinoyl)benzoate

  • MDL number MFCD07699460
  • Cas Number : 898785-81-4
  • Molecular Weight : 271.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-[(2-methoxy-3-pyridinyl)carbonyl]benzoate
Inchi Code 1S/C15H13NO4/c1-19-14-12(8-5-9-16-14)13(17)10-6-3-4-7-11(10)15(18)20-2/h3-9H,1-2H3
Inchi Key QKOGXMPAENSDPH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5577

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5577
Availability details for
Quantity
  • Availability
  • Add to Cart