RIE156322479 Rieke Metals, Inc.

Methyl 3-(2-methoxynicotinoyl)benzoate

  • MDL number MFCD07699461
  • Cas Number : 898785-83-6
  • Molecular Weight : 271.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5578

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5578
Availability details for
Quantity
  • Availability
  • Add to Cart