RIE156322484 Rieke Metals, Inc.

2-Methoxy-3-(3-cyanophenylacetyl)pyridine

  • MDL number MFCD07699466
  • Cas Number : 898785-93-8
  • Molecular Weight : 252.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5583

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5583
Availability details for
Quantity
  • Availability
  • Add to Cart