RIE156322485 Rieke Metals, Inc.

2-Methoxy-3-(4-cyanophenylacetyl)pyridine

  • MDL number MFCD07783006
  • Cas Number : 898785-95-0
  • Molecular Weight : 238.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5584

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5584
Availability details for
Quantity
  • Availability
  • Add to Cart