RIE156322488 Rieke Metals, Inc.

2-Methoxy-5-picolinoylpyridine

  • MDL number MFCD07699470
  • Cas Number : 381725-44-6
  • Molecular Weight : 214.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5587

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5587
Availability details for
Quantity
  • Availability
  • Add to Cart