RIE156322489 Rieke Metals, Inc.

2-Methoxy-3-picolinoylpyridine

  • MDL number MFCD07699471
  • Cas Number : 898786-01-1
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methoxy-3-pyridinyl)(2-pyridinyl)methanone
Inchi Code 1S/C12H10N2O2/c1-16-12-9(5-4-8-14-12)11(15)10-6-2-3-7-13-10/h2-8H,1H3
Inchi Key FSIZLYHTSHDMAL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5588

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5588
Availability details for
Quantity
  • Availability
  • Add to Cart