RIE156322491 Rieke Metals, Inc.

Methyl 3-(6-methoxynicotinoyl)benzoate

  • MDL number MFCD07699473
  • Cas Number : 898786-05-5
  • Molecular Weight : 271.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5590

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5590
Availability details for
Quantity
  • Availability
  • Add to Cart