RIE156322493 Rieke Metals, Inc.

2-Methoxy-5-(3-methyl-2-thenoyl)pyridine

  • MDL number MFCD07699475
  • Cas Number : 898786-11-3
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methoxy-3-pyridinyl)(3-methyl-2-thienyl)methanone
Inchi Code 1S/C12H11NO2S/c1-8-5-6-16-12(8)11(14)9-3-4-10(15-2)13-7-9/h3-7H,1-2H3
Inchi Key NLHCKFIXIFVLBH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5592

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5592
Availability details for
Quantity
  • Availability
  • Add to Cart