RIE156322494 Rieke Metals, Inc.

2-Methoxy-3-thenoylpyridine

  • MDL number MFCD07699476
  • Cas Number : 332134-11-9
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5593

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5593
Availability details for
Quantity
  • Availability
  • Add to Cart