RIE156322495 Rieke Metals, Inc.

2-Methoxy-5-thenoylpyridine

  • MDL number MFCD07699477
  • Cas Number : 898786-14-6
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5594

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5594
Availability details for
Quantity
  • Availability
  • Add to Cart