RIE156322496 Rieke Metals, Inc.

Methyl 5-(2-methoxynicotinoyl) thiophene-2-carboxylate

  • MDL number MFCD07699478
  • Cas Number : 898786-17-9
  • Molecular Weight : 277.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5595

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5595
Availability details for
Quantity
  • Availability
  • Add to Cart