RIE156322498 Rieke Metals, Inc.

Methyl 5-(2-methoxynicotinoyl)-2- furanoate

  • MDL number MFCD07699480
  • Cas Number : 898786-23-7
  • Molecular Weight : 261.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5597

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5597
Availability details for
Quantity
  • Availability
  • Add to Cart