RIE156322499 Rieke Metals, Inc.

Methyl 5-(6-methoxynicotinoyl)-2- furanoate

  • MDL number MFCD07699481
  • Cas Number : 898786-26-0
  • Molecular Weight : 261.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5598

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5598
Availability details for
Quantity
  • Availability
  • Add to Cart