RIE156322500 Rieke Metals, Inc.

3-(2-Furoyl)-2-methoxypyridine

  • MDL number MFCD07699482
  • Cas Number : 898786-29-3
  • Molecular Weight : 203.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5599

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5599
Availability details for
Quantity
  • Availability
  • Add to Cart