RIE156322501 Rieke Metals, Inc.

5-(2-Furoyl)-2-methoxypyridine

  • MDL number MFCD07699483
  • Cas Number : 898786-32-8
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-furyl(6-methoxy-3-pyridinyl)methanone
Inchi Code 1S/C11H9NO3/c1-14-10-5-4-8(7-12-10)11(13)9-3-2-6-15-9/h2-7H,1H3
Inchi Key QUMUNJFKCHRNIO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5600

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5600
Availability details for
Quantity
  • Availability
  • Add to Cart