RIE156322505 Rieke Metals, Inc.

5-(2-Acetoxybenzoyl)-2-chloropyridine

  • MDL number MFCD07699487
  • Cas Number : 898786-44-2
  • Molecular Weight : 275.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5602-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5602-1
Availability details for
Quantity
  • Availability
  • Add to Cart