RIE156322506 Rieke Metals, Inc.

5-(3-Acetoxybenzoyl)-2-chloropyridine

  • MDL number MFCD07699488
  • Cas Number : 898786-47-5
  • Molecular Weight : 275.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5602-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5602-2
Availability details for
Quantity
  • Availability
  • Add to Cart