RIE156322507 Rieke Metals, Inc.

5-(4-Acetoxybenzoyl)-2-chloropyridine

  • MDL number MFCD07699489
  • Cas Number : 898786-50-0
  • Molecular Weight : 275.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5602-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5602-3
Availability details for
Quantity
  • Availability
  • Add to Cart