RIE156322511 Rieke Metals, Inc.

2-(2-Acetoxybenzoyl) furan

  • MDL number MFCD07699493
  • Cas Number : 898766-17-1
  • Molecular Weight : 230.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5604-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5604-1
Availability details for
Quantity
  • Availability
  • Add to Cart