RIE156322516 Rieke Metals, Inc.

3-(4-Acetoxybenzoyl)-5-bromopyridine

  • MDL number MFCD07699498
  • Cas Number : 898766-32-0
  • Molecular Weight : 320.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5605-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5605-3
Availability details for
Quantity
  • Availability
  • Add to Cart