RIE156322532 Rieke Metals, Inc.

7-Oxo-7-phenylheptanenitrile

  • MDL number MFCD02260533
  • Cas Number : 39755-14-1
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6001E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6001E
Availability details for
Quantity
  • Availability
  • Add to Cart