RIE156322535 Rieke Metals, Inc.

7-(3-bromophenyl)-7-oxoheptanenitrile

  • MDL number MFCD02260541
  • Cas Number : 898766-82-0
  • Molecular Weight : 280.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(3-bromophenyl)-7-oxoheptanenitrile
Inchi Code 1S/C13H14BrNO/c14-12-7-5-6-11(10-12)13(16)8-3-1-2-4-9-15/h5-7,10H,1-4,8H2
Inchi Key HOMPSBVEEFFHKC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6003e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6003e
Availability details for
Quantity
  • Availability
  • Add to Cart