RIE156322536 Rieke Metals, Inc.

8-(3-bromophenyl)-8-oxooctanenitrile

  • MDL number MFCD02260542
  • Cas Number : 898766-84-2
  • Molecular Weight : 294.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6003F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6003F
Availability details for
Quantity
  • Availability
  • Add to Cart