RIE156322537 Rieke Metals, Inc.

5-(4-bromophenyl)-5-oxovaleronitrile

  • MDL number MFCD00672045
  • Cas Number : 898766-86-4
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-bromophenyl)-5-oxopentanenitrile
Inchi Code 1S/C11H10BrNO/c12-10-6-4-9(5-7-10)11(14)3-1-2-8-13/h4-7H,1-3H2
Inchi Key YHGHWUIHGFVXGS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6004c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6004c
Availability details for
Quantity
  • Availability
  • Add to Cart