RIE156322539 Rieke Metals, Inc.

8-(4-bromophenyl)-8-oxooctanenitrile

  • MDL number MFCD02260546
  • Cas Number : 898766-90-0
  • Molecular Weight : 294.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-(4-bromophenyl)-8-oxooctanenitrile
Inchi Code 1S/C14H16BrNO/c15-13-9-7-12(8-10-13)14(17)6-4-2-1-3-5-11-16/h7-10H,1-6H2
Inchi Key MOBLJGZBZZWAJD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6004f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6004f
Availability details for
Quantity
  • Availability
  • Add to Cart