RIE156322540 Rieke Metals, Inc.

1-(2-bromophenyl)-5-chloro-1-oxopentane

  • MDL number MFCD00672050
  • Cas Number : 487058-92-4
  • Molecular Weight : 275.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromophenyl)-5-chloro-1-pentanone
Inchi Code 1S/C11H12BrClO/c12-10-6-2-1-5-9(10)11(14)7-3-4-8-13/h1-2,5-6H,3-4,7-8H2
Inchi Key AZMKKIHXEPLPGT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6008d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6008d
Availability details for
Quantity
  • Availability
  • Add to Cart