RIE156322542 Rieke Metals, Inc.

1-(2-bromophenyl)-7-chloro-1-oxoheptane

  • MDL number MFCD00672048
  • Cas Number : 898766-94-4
  • Molecular Weight : 303.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6008F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6008F
Availability details for
Quantity
  • Availability
  • Add to Cart