RIE156322543 Rieke Metals, Inc.

1-(3-Bromophenyl)-4-chloro-1-oxobutane

  • MDL number MFCD02260550
  • Cas Number : 494863-34-2
  • Molecular Weight : 261.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromophenyl)-4-chloro-1-butanone
Inchi Code 1S/C10H10BrClO/c11-9-4-1-3-8(7-9)10(13)5-2-6-12/h1,3-4,7H,2,5-6H2
Inchi Key AYCQFQSAPFTAFF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6011c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6011c
Availability details for
Quantity
  • Availability
  • Add to Cart