RIE156322544 Rieke Metals, Inc.

1-(3-bromophenyl)-5-chloro-1-oxopentane

  • MDL number MFCD00672053
  • Cas Number : 487058-88-8
  • Molecular Weight : 275.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6011D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6011D
Availability details for
Quantity
  • Availability
  • Add to Cart