RIE156322545 Rieke Metals, Inc.

1-(3-bromophenyl)-6-chloro-1-oxohexane

  • MDL number MFCD00672052
  • Cas Number : 898766-96-6
  • Molecular Weight : 289.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6011E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6011E
Availability details for
Quantity
  • Availability
  • Add to Cart