RIE156322546 Rieke Metals, Inc.

1-(3-bromophenyl)-7-chloro-1-oxoheptane

  • MDL number MFCD00672051
  • Cas Number : 898766-98-8
  • Molecular Weight : 303.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6011F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6011F
Availability details for
Quantity
  • Availability
  • Add to Cart