RIE156322547 Rieke Metals, Inc.

1-(4-bromophenyl)-5-chloro-1-oxopentane

  • MDL number MFCD02260551
  • Cas Number : 54874-12-3
  • Molecular Weight : 275.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-5-chloro-1-pentanone
Inchi Code 1S/C11H12BrClO/c12-10-6-4-9(5-7-10)11(14)3-1-2-8-13/h4-7H,1-3,8H2
Inchi Key UNIORYRWTPWEAI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6014d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6014d
Availability details for
Quantity
  • Availability
  • Add to Cart