RIE156322548 Rieke Metals, Inc.

1-(4-bromophenyl)-6-chloro-1-oxohexane

  • MDL number MFCD00672055
  • Cas Number : 898767-00-5
  • Molecular Weight : 289.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6014e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6014e
Availability details for
Quantity
  • Availability
  • Add to Cart