RIE156322549 Rieke Metals, Inc.

1-(4-bromophenyl)-7-chloro-1-oxoheptane

  • MDL number MFCD00672054
  • Cas Number : 193065-67-7
  • Molecular Weight : 303.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-7-chloro-1-heptanone
Inchi Code 1S/C13H16BrClO/c14-12-8-6-11(7-9-12)13(16)5-3-1-2-4-10-15/h6-9H,1-5,10H2
Inchi Key RGPPQQUQFFSDQO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6014f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6014f
Availability details for
Quantity
  • Availability
  • Add to Cart