RIE156322550 Rieke Metals, Inc.

5-Chloro-1-(4-fluorophenyl)-1-oxopentane

  • MDL number MFCD01319941
  • Cas Number : 17135-46-5
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6016D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6016D
Availability details for
Quantity
  • Availability
  • Add to Cart