RIE156322551 Rieke Metals, Inc.

3-Chloro-1-(3-fluorophenyl)-1-oxopropane

  • MDL number MFCD07699514
  • Cas Number : 83406-26-2
  • Molecular Weight : 186.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-1-(3-fluorophenyl)-1-propanone
Inchi Code 1S/C9H8ClFO/c10-5-4-9(12)7-2-1-3-8(11)6-7/h1-3,6H,4-5H2
Inchi Key SOMLWYGQLWEMCT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6017b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6017b
Availability details for
Quantity
  • Availability
  • Add to Cart