RIE156322552 Rieke Metals, Inc.

4-Chloro-1-(3-fluorophenyl)-1-oxobutane

  • MDL number MFCD02260552
  • Cas Number : 3110-52-9
  • Molecular Weight : 200.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6017C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6017C
Availability details for
Quantity
  • Availability
  • Add to Cart