RIE156322554 Rieke Metals, Inc.

6-chloro-1-(3-fluorophenyl)-1-oxohexane

  • MDL number MFCD01319945
  • Cas Number : 488098-58-4
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6017E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6017E
Availability details for
Quantity
  • Availability
  • Add to Cart