RIE156322557 Rieke Metals, Inc.

4-Chloro-1-(2-fluorophenyl)-1-oxobutane

  • MDL number MFCD02260553
  • Cas Number : 2823-19-0
  • Molecular Weight : 200.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6018C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6018C
Availability details for
Quantity
  • Availability
  • Add to Cart