RIE156322558 Rieke Metals, Inc.

5-chloro-1-(2-fluorophenyl)-1-oxopentane

  • MDL number MFCD01319947
  • Cas Number : 487058-75-3
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6018D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6018D
Availability details for
Quantity
  • Availability
  • Add to Cart