RIE156322559 Rieke Metals, Inc.

6-chloro-1-(2-fluorophenyl)-1-oxohexane

  • MDL number MFCD01319948
  • Cas Number : 488098-59-5
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6018E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6018E
Availability details for
Quantity
  • Availability
  • Add to Cart