RIE156322560 Rieke Metals, Inc.

7-chloro-1-(2-fluorophenyl)-1-oxoheptane

  • MDL number MFCD01319949
  • Cas Number : 487058-11-7
  • Molecular Weight : 242.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6018F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6018F
Availability details for
Quantity
  • Availability
  • Add to Cart