RIE156322561 Rieke Metals, Inc.

5-(4-Fluorophenyl)-5-oxovaleronitrile

  • MDL number MFCD01319951
  • Cas Number : 898767-06-1
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-fluorophenyl)-5-oxopentanenitrile
Inchi Code 1S/C11H10FNO/c12-10-6-4-9(5-7-10)11(14)3-1-2-8-13/h4-7H,1-3H2
Inchi Key CDGYAXKSYRIFEX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6019c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6019c
Availability details for
Quantity
  • Availability
  • Add to Cart