RIE156322562 Rieke Metals, Inc.

6-(4-fluorophenyl)-6-oxohexanenitrile

  • MDL number MFCD01319952
  • Cas Number : 898767-09-4
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6019D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6019D
Availability details for
Quantity
  • Availability
  • Add to Cart