RIE156322565 Rieke Metals, Inc.

4-(3-Fluorophenyl)-4-oxobutyronitrile

  • MDL number MFCD01319953
  • Cas Number : 298690-71-8
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6020B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6020B
Availability details for
Quantity
  • Availability
  • Add to Cart