RIE156322566 Rieke Metals, Inc.

5-(3-fluorophenyl)-5-oxovaleronitrile

  • MDL number MFCD01319954
  • Cas Number : 898767-18-5
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6020C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6020C
Availability details for
Quantity
  • Availability
  • Add to Cart