RIE156322570 Rieke Metals, Inc.

4-(2-Fluorophenyl)-4-oxobutyronitrile

  • MDL number MFCD01319956
  • Cas Number : 898767-30-1
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-fluorophenyl)-4-oxobutanenitrile
Inchi Code 1S/C10H8FNO/c11-9-5-2-1-4-8(9)10(13)6-3-7-12/h1-2,4-5H,3,6H2
Inchi Key GGERJUSCKQINCU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6021b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6021b
Availability details for
Quantity
  • Availability
  • Add to Cart