RIE156322571 Rieke Metals, Inc.

5-(2-Fluorophenyl)-5-oxovaleronitrile

  • MDL number MFCD01319957
  • Cas Number : 898767-33-4
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6021C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6021C
Availability details for
Quantity
  • Availability
  • Add to Cart